Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306229.3 | NP_001293158.1 | 2 - 3 | 213 | 101 | |
| NM_001306310.3 | NP_001293239.1 | 41 - 42 | 9897 | 101 | ||
| NM_001306311.3 | NP_001293240.1 | 30 - 31 | 7098 | 101 | ||
| NM_001306312.3 | NP_001293241.1 | 18 - 19 | 4284 | 101 | ||
| NM_001306313.4 | NP_001293242.1 | 44 - 45 | 10542 | 101 | ||
| NM_001306314.3 | NP_001293243.1 | 40 - 41 | 9864 | 101 | ||
| NM_001383731.1 | 40 - 41 | 9915 | 101 | |||
| NM_060545.6 | NP_492946.1 | 45 - 46 | 10637 | 101 | ||
| NM_060546.7 | NP_492947.1 | 3 - 4 | 526 | 101 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map