Assay Details
| Gene Symbol: dys-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306229.3 | NP_001293158.1 | 3 - 4 | 340 | 76 | |
| NM_001306230.3 | NP_001293159.1 | 1 - 2 | 127 | 76 | ||
| NM_001306310.3 | NP_001293239.1 | 42 - 43 | 10024 | 76 | ||
| NM_001306311.3 | NP_001293240.1 | 31 - 32 | 7225 | 76 | ||
| NM_001306312.3 | NP_001293241.1 | 19 - 20 | 4411 | 76 | ||
| NM_001306313.4 | NP_001293242.1 | 45 - 46 | 10669 | 76 | ||
| NM_001306314.3 | NP_001293243.1 | 41 - 42 | 9991 | 76 | ||
| NM_001383731.1 | 41 - 42 | 10042 | 76 | |||
| NM_060545.6 | NP_492946.1 | 46 - 47 | 10764 | 76 | ||
| NM_060546.7 | NP_492947.1 | 4 - 5 | 653 | 76 | ||
More Information
Set Membership: |
|

dys-1
Genomic Map