Assay Details
| Gene Symbol: spe-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060554.5 | NP_492955.1 | 2 - 3 | 163 | 144 | |
| GenBank mRNA | AF049327.1 | - | 2 - 3 | 141 | 144 | |
More Information
Set Membership: |
|

spe-9
Genomic Map