Assay Details
| Gene Symbol: kpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060573.6 | NP_492974.2 | 2 - 3 | 656 | 66 | |
| NR_132618.1 | 2 - 3 | 556 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kpc-1| Gene Symbol: kpc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060573.6 | NP_492974.2 | 2 - 3 | 656 | 66 | |
| NR_132618.1 | 2 - 3 | 556 | 66 | |||
Set Membership: |
|