Assay Details
| Gene Symbol: rga-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060634.7 | NP_493035.1 | 7 - 8 | 1559 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rga-2| Gene Symbol: rga-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060634.7 | NP_493035.1 | 7 - 8 | 1559 | 68 | |
Set Membership: |
|