Assay Details
| Gene Symbol: tmem-189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060635.8 | NP_493036.1 | 5 - 6 | 794 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tmem-189| Gene Symbol: tmem-189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060635.8 | NP_493036.1 | 5 - 6 | 794 | 104 | |
Set Membership: |
|