Assay Details
| Gene Symbol: Y47H10A.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060652.4 | NP_493053.2 | 5 - 6 | 1021 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y47H10A.2| Gene Symbol: Y47H10A.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060652.4 | NP_493053.2 | 5 - 6 | 1021 | 155 | |
Set Membership: |
|