Assay Details
| Gene Symbol: sri-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060670.2 | NP_493071.2 | 2 - 3 | 369 | 197 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-16| Gene Symbol: sri-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060670.2 | NP_493071.2 | 2 - 3 | 369 | 197 | |
Set Membership: |
|