Assay Details
| Gene Symbol: T15D6.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060740.3 | NP_493141.1 | 2 - 3 | 239 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T15D6.10| Gene Symbol: T15D6.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060740.3 | NP_493141.1 | 2 - 3 | 239 | 121 | |
Set Membership: |
|