Assay Details
| Gene Symbol: T15D6.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060742.2 | NP_493143.1 | 4 - 5 | 984 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T15D6.12| Gene Symbol: T15D6.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060742.2 | NP_493143.1 | 4 - 5 | 984 | 87 | |
Set Membership: |
|