Assay Details
| Gene Symbol: pri-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264993.2 | NP_001251922.1 | 3 - 4 | 764 | 102 | |
| NM_001264994.3 | NP_001251923.1 | 1 - 2 | 213 | 102 | ||
More Information
Set Membership: |
|

pri-2
Genomic Map