Assay Details
| Gene Symbol: pri-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264993.2 | NP_001251922.1 | 4 - 5 | 1021 | 71 | |
| NM_001264994.3 | NP_001251923.1 | 2 - 3 | 470 | 71 | ||
More Information
Set Membership: |
|

pri-2
Genomic Map