Assay Details
| Gene Symbol: hmg-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060777.6 | NP_493178.1 | 5 - 6 | 1039 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hmg-20| Gene Symbol: hmg-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060777.6 | NP_493178.1 | 5 - 6 | 1039 | 105 | |
Set Membership: |
|