Assay Details
| Gene Symbol: dlk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026272.4 | NP_001021443.1 | 4 - 5 | 859 | 138 | |
| NM_001026273.6 | NP_001021444.1 | 4 - 5 | 859 | 138 | ||
| NM_001026274.5 | NP_001021445.1 | 4 - 5 | 854 | 138 | ||
| NM_001136297.4 | NP_001129769.1 | 2 - 3 | 635 | 138 | ||
More Information
Set Membership: |
|

dlk-1
Genomic Map