Assay Details
| Gene Symbol: dlk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026272.4 | NP_001021443.1 | 10 - 11 | 2700 | 62 | |
| NM_001026273.6 | NP_001021444.1 | 10 - 11 | 2679 | 62 | ||
| NM_001136297.4 | NP_001129769.1 | 8 - 9 | 2476 | 62 | ||
| NM_001265008.3 | NP_001251937.1 | 1 - 2 | 532 | 62 | ||
More Information
Set Membership: |
|

dlk-1
Genomic Map