Assay Details
| Gene Symbol: uba-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306225.3 | NP_001293154.1 | 4 - 5 | 451 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

uba-2| Gene Symbol: uba-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306225.3 | NP_001293154.1 | 4 - 5 | 451 | 65 | |
Set Membership: |
|