Assay Details
| Gene Symbol: ptr-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377664.1 | NP_001364498.1 | 11 - 12 | 1972 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-17| Gene Symbol: ptr-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377664.1 | NP_001364498.1 | 11 - 12 | 1972 | 141 | |
Set Membership: |
|