Assay Details
| Gene Symbol: gale-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025880.3 | NP_001021051.1 | 2 - 3 | 168 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gale-1| Gene Symbol: gale-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025880.3 | NP_001021051.1 | 2 - 3 | 168 | 108 | |
Set Membership: |
|