Assay Details
| Gene Symbol: gale-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025880.3 | NP_001021051.1 | 4 - 5 | 831 | 138 | |
| NM_001025881.6 | NP_001021052.1 | 4 - 5 | 839 | 138 | ||
More Information
Set Membership: |
|

gale-1
Genomic Map