Assay Details
| Gene Symbol: pals-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060895.4 | NP_493296.2 | 3 - 4 | 318 | 80 | |
| NR_132565.1 | 2 - 3 | 244 | 80 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-3| Gene Symbol: pals-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060895.4 | NP_493296.2 | 3 - 4 | 318 | 80 | |
| NR_132565.1 | 2 - 3 | 244 | 80 | |||
Set Membership: |
|