Assay Details
| Gene Symbol: pals-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047172.1 | NP_001040637.1 | 1 - 2 | 212 | 65 | |
| Gene Symbol: pals-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132565.1 | 5 - 6 | 1082 | 65 | ||
More Information
Set Membership: |
|

pals-12
Genomic Map