Assay Details
| Gene Symbol: scrm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060920.2 | NP_493321.1 | 1 - 2 | 386 | 74 | |
| GenBank mRNA | EF445110.1 | - | 1 - 2 | 367 | 74 | |
More Information
Set Membership: |
|

scrm-2
Genomic Map