Assay Details
| Gene Symbol: spr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060965.6 | NP_493366.1 | 1 - 2 | 88 | 90 | |
| GenBank mRNA | AX799789.1 | - | 1 - 2 | 86 | 90 | |
| AY152852.1 | - | 1 - 2 | 86 | 90 | ||
More Information
Set Membership: |
|

spr-5
Genomic Map