Assay Details
| Gene Symbol: cyn-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060977.5 | NP_493378.1 | 4 - 5 | 1798 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyn-15| Gene Symbol: cyn-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060977.5 | NP_493378.1 | 4 - 5 | 1798 | 106 | |
Set Membership: |
|