Assay Details
Gene Symbol: nyn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383746.1 | 1 - 2 | 597 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
