Assay Details
| Gene Symbol: ints-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060995.8 | NP_493396.1 | 5 - 6 | 1634 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ints-4| Gene Symbol: ints-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060995.8 | NP_493396.1 | 5 - 6 | 1634 | 57 | |
Set Membership: |
|