Assay Details
| Gene Symbol: mys-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322689.3 | NP_001309531.1 | 1 - 2 | 164 | 117 | |
| NM_001330941.4 | NP_001317768.1 | 1 - 2 | 166 | 117 | ||
More Information
Set Membership: |
|

mys-2
Genomic Map