Assay Details
| Gene Symbol: mys-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322689.3 | NP_001309531.1 | 9 - 10 | 1523 | 114 | |
| NM_001330942.3 | NP_001317769.1 | 2 - 3 | 456 | 114 | ||
More Information
Set Membership: |
|

mys-2
Genomic Map