Assay Details
| Gene Symbol: pry-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061073.6 | NP_493474.2 | 5 - 6 | 583 | 59 | |
| GenBank mRNA | AF468834.1 | - | 5 - 6 | 585 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pry-1| Gene Symbol: pry-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061073.6 | NP_493474.2 | 5 - 6 | 583 | 59 | |
| GenBank mRNA | AF468834.1 | - | 5 - 6 | 585 | 59 | |
Set Membership: |
|