Assay Details
| Gene Symbol: pry-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061073.6 | NP_493474.2 | 9 - 10 | 1550 | 104 | |
| GenBank mRNA | AF468834.1 | - | 9 - 10 | 1552 | 104 | |
More Information
Set Membership: |
|

pry-1
Genomic Map