Assay Details
| Gene Symbol: ptr-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061202.6 | NP_493603.1 | 12 - 13 | 3002 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-23| Gene Symbol: ptr-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061202.6 | NP_493603.1 | 12 - 13 | 3002 | 63 | |
Set Membership: |
|