Assay Details
| Gene Symbol: pole-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061215.5 | NP_493616.1 | 7 - 8 | 2368 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pole-1| Gene Symbol: pole-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061215.5 | NP_493616.1 | 7 - 8 | 2368 | 71 | |
Set Membership: |
|