Assay Details
| Gene Symbol: F33H2.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061216.9 | NP_493617.2 | 2 - 3 | 414 | 169 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F33H2.6| Gene Symbol: F33H2.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061216.9 | NP_493617.2 | 2 - 3 | 414 | 169 | |
Set Membership: |
|