Assay Details
| Gene Symbol: rpt-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026942.5 | NP_001022113.1 | 2 - 3 | 463 | 101 | |
| NM_001026943.4 | NP_001022114.1 | 1 - 2 | 358 | 101 | ||
More Information
Set Membership: |
|

rpt-4
Genomic Map