Assay Details
| Gene Symbol: ascc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129119.3 | NP_001122591.1 | 1 - 2 | 179 | 59 | |
| NR_132568.1 | 1 - 2 | 177 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ascc-1| Gene Symbol: ascc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129119.3 | NP_001122591.1 | 1 - 2 | 179 | 59 | |
| NR_132568.1 | 1 - 2 | 177 | 59 | |||
Set Membership: |
|