Assay Details
| Gene Symbol: ascc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129119.3 | NP_001122591.1 | 6 - 7 | 756 | 68 | |
| NR_132568.1 | 6 - 7 | 754 | 68 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ascc-1| Gene Symbol: ascc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129119.3 | NP_001122591.1 | 6 - 7 | 756 | 68 | |
| NR_132568.1 | 6 - 7 | 754 | 68 | |||
Set Membership: |
|