Assay Details
| Gene Symbol: nmur-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061265.4 | NP_493666.2 | 1 - 2 | 81 | 168 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nmur-2| Gene Symbol: nmur-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061265.4 | NP_493666.2 | 1 - 2 | 81 | 168 | |
Set Membership: |
|