Assay Details
| Gene Symbol: etr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381290.2 | NP_001367867.1 | 8 - 9 | 1317 | 54 | |
| NM_001381294.1 | NP_001367872.1 | 8 - 9 | 1292 | 54 | ||
| NM_001381298.1 | NP_001368524.1 | 6 - 7 | 670 | 54 | ||
| NM_001381301.1 | NP_001368527.1 | 4 - 5 | 544 | 54 | ||
| NM_001381302.1 | NP_001367869.1 | 4 - 5 | 559 | 54 | ||
| NM_001381305.1 | NP_001368530.1 | 3 - 4 | 346 | 54 | ||
| NM_001381308.1 | NP_001368533.1 | 1 - 2 | 118 | 54 | ||
| NR_132573.1 | 8 - 9 | 1255 | 54 | |||
| GenBank mRNA | EF523766.1 | - | 8 - 9 | 1255 | 54 | |
| EF523770.1 | - | 7 - 8 | 1042 | 54 | ||
| U53931.1 | - | 8 - 9 | 1318 | 54 | ||
More Information
Set Membership: |
|

etr-1
Genomic Map