Assay Details
| Gene Symbol: chs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061281.6 | NP_493682.2 | 1 - 2 | 476 | 110 | |
| GenBank mRNA | AY874872.1 | - | 1 - 2 | 252 | 110 | |
More Information
Set Membership: |
|

chs-2
Genomic Map