Assay Details
| Gene Symbol: chs-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313615.3 | NP_001300544.1 | 7 - 8 | 1499 | 92 | |
| NM_001313616.3 | NP_001300545.1 | 9 - 10 | 1781 | 92 | ||
| NM_061281.6 | NP_493682.2 | 7 - 8 | 1909 | 92 | ||
| GenBank mRNA | AY874872.1 | - | 7 - 8 | 1685 | 92 | |
More Information
Set Membership: |
|

chs-2
Genomic Map