Assay Details
| Gene Symbol: ptr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061317.7 | NP_493718.2 | 11 - 12 | 2119 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-3| Gene Symbol: ptr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061317.7 | NP_493718.2 | 11 - 12 | 2119 | 75 | |
Set Membership: |
|