Assay Details
| Gene Symbol: nfyb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316872.3 | NP_001303801.1 | 3 - 4 | 462 | 117 | |
| NM_061339.6 | NP_493740.1 | 4 - 5 | 713 | 117 | ||
More Information
Set Membership: |
|

nfyb-1
Genomic Map