Assay Details
| Gene Symbol: mlt-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061354.5 | NP_493755.1 | 4 - 5 | 1123 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mlt-10| Gene Symbol: mlt-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061354.5 | NP_493755.1 | 4 - 5 | 1123 | 85 | |
Set Membership: |
|