Assay Details
| Gene Symbol: mlt-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061354.5 | NP_493755.1 | 7 - 8 | 1605 | 73 | |
| Gene Symbol: T27A1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316855.2 | NP_001303784.1 | 2651 | 73 | ||
More Information
Set Membership: |
|

mlt-10
Genomic Map