Assay Details
| Gene Symbol: cdc-37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381317.1 | NP_001367479.1 | 3 - 4 | 1082 | 80 | |
| NM_001381318.1 | NP_001367478.1 | 3 - 4 | 1037 | 80 | ||
More Information
Set Membership: |
|

cdc-37
Genomic Map