Assay Details
| Gene Symbol: fbxb-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061362.3 | NP_493763.1 | 1 | 577 | 75 | |
| Gene Symbol: W08F4.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133623.1 | 4 | 1501 | 75 | ||
More Information
Set Membership: |
|

fbxb-14
Genomic Map