Assay Details
| Gene Symbol: sri-46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316857.3 | NP_001303786.1 | 1 - 2 | 164 | 84 | |
| NM_061384.2 | NP_493785.1 | 3 - 4 | 545 | 84 | ||
More Information
Set Membership: |
|

sri-46
Genomic Map