Assay Details
| Gene Symbol: thoc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061395.3 | NP_493796.2 | 9 - 10 | 1019 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

thoc-1| Gene Symbol: thoc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061395.3 | NP_493796.2 | 9 - 10 | 1019 | 138 | |
Set Membership: |
|