Assay Details
Gene Symbol: lmd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380707.1 | NP_001367831.1 | 1 - 2 | 61 | 83 | |
NM_072772.4 | NP_505173.2 | 1 - 2 | 61 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
