Assay Details
Gene Symbol: lmd-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373062.3 | NP_001360511.1 | 6 - 7 | 1193 | 71 | |
NM_001373063.2 | NP_001360537.1 | 5 - 6 | 1160 | 71 | ||
NM_001373064.1 | NP_001360538.1 | 5 - 6 | 1160 | 71 | ||
NM_001380707.1 | NP_001367831.1 | 6 - 7 | 1223 | 71 | ||
NM_072772.4 | NP_505173.2 | 6 - 7 | 1223 | 71 | ||
NM_072773.7 | NP_505174.3 | 6 - 7 | 1229 | 71 | ||
NM_072774.9 | NP_505175.2 | 6 - 7 | 1229 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_072774.9 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
